2-(tert-Butylsulphanyl)aniline 97%

Lieferant: Apollo Scientific

Synonyme: 2-(tert-Butylthio)aniline

OR962840-1G OR962840-5G
APOSOR962840-1GEA 97.3 EUR
APOSOR962840-1G APOSOR962840-5G
2-(tert-Butylsulphanyl)aniline 97%
2-(tert-Butylsulphanyl)aniline
Formel: C₁₀H₁₅NS
Molecular Weight: 181,298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11136099
CAS-Nummer: 51942-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR