2-(Pentan-3-yloxy)acetic acid 98%

Lieferant: Apollo Scientific

OR962845-1G OR962845-5G
APOSOR962845-1GEA 248 EUR
APOSOR962845-1G APOSOR962845-5G
2-(Pentan-3-yloxy)acetic acid 98%
2-(Pentan-3-yloxy)acetic acid
Formel: C₇H₁₄O₃
Molecular Weight: 146,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520462
CAS-Nummer: 189955-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR