4-Bromo-3-methylpyridine 1-oxide 97%

Lieferant: Apollo Scientific

OR962889-1G OR962889-5G
APOSOR962889-1GEA 248 EUR
APOSOR962889-1G APOSOR962889-5G
4-Bromo-3-methylpyridine 1-oxide 97%
4-Bromo-3-methylpyridine 1-oxide
Formel: C₆H₆BrNO
Molecular Weight: 188.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870743
CAS-Nummer: 10168-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR