(2R)-2-Methylpyrrolidine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-R-proline, (R)-2-Methylpyrrolidine-2-carboxylic acid, H-α-Me-D-Pro-OH

OR962960-1G
APOSOR962960-1GEA 66.7 EUR
APOSOR962960-1G
(2R)-2-Methylpyrrolidine-2-carboxylic acid 95%
(2R)-2-Methylpyrrolidine-2-carboxylic acid
Formel: C₆H₁₁NO₂
Molecular Weight: 129,157 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08274534
CAS-Nummer: 63399-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR