2-(N-Boc-methylamino)acetonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (cyanomethyl)(methyl)carbamate
OR963413-10G
OR963413-1G
OR963413-5G
APOSOR963413-10GEA
119
EUR
APOSOR963413-10G
APOSOR963413-1G
APOSOR963413-5G
2-(N-Boc-methylamino)acetonitrile 95%
2-(N-Boc-methylamino)acetonitrile
Formel:
C₈H₁₄N₂O₂ Molecular Weight: 170,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658361 CAS-Nummer: 180976-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...