2-(Pyrimidin-2-yloxy)benzonitrile 95%

Lieferant: Apollo Scientific

OR963445-1G OR963445-250MG
APOSOR963445-1GEA 342 EUR
APOSOR963445-1G APOSOR963445-250MG
2-(Pyrimidin-2-yloxy)benzonitrile 95%
2-(Pyrimidin-2-yloxy)benzonitrile
Formel: C₁₁H₇N₃O
Molecular Weight: 197,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12068354
CAS-Nummer: 1159822-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR