2,4-Dimethyloxazole-5-carboxaldehyde 95%
Lieferant: Apollo Scientific
OR963593-1G
OR963593-250MG
APOSOR963593-1GEA
770
EUR
APOSOR963593-1G
APOSOR963593-250MG
2,4-Dimethyloxazole-5-carboxaldehyde 95%
2,4-Dimethyloxazole-5-carboxaldehyde
Formel:
C₆H₇NO₂ Molecular Weight: 125,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738688 CAS-Nummer: 69062-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...