2-Amino-6-phenylpyridine 97%

Lieferant: Apollo Scientific

OR963733-10G OR963733-1G OR963733-5G
APOSOR963733-10GEA 463 EUR
APOSOR963733-10G APOSOR963733-1G APOSOR963733-5G
2-Amino-6-phenylpyridine 97%
2-Amino-6-phenylpyridine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234719
CAS-Nummer: 39774-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR