2-Chloro-1-(3-chloro-phenyl)-ethanol 95%

Lieferant: Apollo Scientific

OR963828-1G OR963828-250MG
APOSOR963828-1GEA 0 EUR
APOSOR963828-1G APOSOR963828-250MG
2-Chloro-1-(3-chloro-phenyl)-ethanol 95%
2-Chloro-1-(3-chloro-phenyl)-ethanol
Formel: C₈H₈Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07784075
CAS-Nummer: 106262-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR