2-Chloro-1-(4-chloro-phenyl)-ethanol 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-1-(4-chlorophenyl)ethanol
OR963832-1G
OR963832-250MG
APOSOR963832-1GEA
91.5
EUR
APOSOR963832-1G
APOSOR963832-250MG
2-Chloro-1-(4-chloro-phenyl)-ethanol 95%
2-Chloro-1-(4-chloro-phenyl)-ethanol
Formel:
C₈H₈Cl₂O Molecular Weight: 191,055 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00462592 CAS-Nummer: 6378-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...