2-Chloro-2-(3-chloro-phenyl)-ethanol 95%

Lieferant: Apollo Scientific

OR963839-1G OR963839-250MG
APOSOR963839-1GEA 647 EUR
APOSOR963839-1G APOSOR963839-250MG
2-Chloro-2-(3-chloro-phenyl)-ethanol 95%
2-Chloro-2-(3-chloro-phenyl)-ethanol
Formel: C₈H₈Cl₂O
Molecular Weight: 191,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784074
CAS-Nummer: 886365-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR