4-Phenoxyphenacyl bromide 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(4-phenoxyphenyl)ethan-1-one

OR9640-1G OR9640-250MG
APOSOR9640-1GEA 401 EUR
APOSOR9640-1G APOSOR9640-250MG
4-Phenoxyphenacyl bromide 90%
4-Phenoxyphenacyl bromide
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291,14 g/mol
Schmelzpunkt: 49…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01419855
CAS-Nummer: 28179-33-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR