2-Methyl-oxazole-5-carboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-methyloxazole-5-carboxylate

OR964036-1G OR964036-250MG
APOSOR964036-1GEA 637 EUR
APOSOR964036-1G APOSOR964036-250MG
2-Methyl-oxazole-5-carboxylic acid methyl ester 95%
2-Methyl-oxazole-5-carboxylic acid methyl ester
Formel: C₆H₇NO₃
Molecular Weight: 141.125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738687
CAS-Nummer: 651059-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR