Methyl 2-phenyloxazole-4-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR964099-1G
APOSOR964099-1GEA
380
EUR
APOSOR964099-1G
Methyl 2-phenyloxazole-4-carboxylate 98%
Methyl 2-phenyloxazole-4-carboxylate
Formel:
C₁₁H₉NO₃ Molecular Weight: 203,194 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00522084 CAS-Nummer: 59171-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...