3,4-Dihydro-4-oxo-7-quinzolinecarboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
4-Oxo-3,4-dihydroquinazoline-7-carboxylic acid
OR964416-1G
OR964416-250MG
APOSOR964416-1GEA
154
EUR
APOSOR964416-1G
APOSOR964416-250MG
3,4-Dihydro-4-oxo-7-quinzolinecarboxylic acid 95%
3,4-Dihydro-4-oxo-7-quinzolinecarboxylic acid
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11983244 CAS-Nummer: 202197-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...