3-Amino-3-methyl-1-pentyne hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methylpent-1-yn-3-amine hydrochloride

OR964685-1G OR964685-5G
APOSOR964685-1GEA 68.3 EUR
APOSOR964685-1G APOSOR964685-5G
3-Amino-3-methyl-1-pentyne hydrochloride 95%
3-Amino-3-methyl-1-pentyne hydrochloride
Formel: C₆H₁₁N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09749829
CAS-Nummer: 108575-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR