5,6,7,8-Tetrahydroisoquinolin-3-amine 97%

Lieferant: Apollo Scientific

OR964708-1G OR964708-250MG
APOSOR964708-1GEA 1060 EUR
APOSOR964708-1G APOSOR964708-250MG
5,6,7,8-Tetrahydroisoquinolin-3-amine 97%
5,6,7,8-Tetrahydroisoquinolin-3-amine
Formel: C₉H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10574676
CAS-Nummer: 69958-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR