1-(3-Pyridinyl)-1-propanamine 95%
Lieferant: Apollo Scientific
Synonyme:
1-(Pyridin-3-yl)propan-1-amine
OR965034-1G
APOSOR965034-1GEA
139
EUR
APOSOR965034-1G
1-(3-Pyridinyl)-1-propanamine 95%
1-(3-Pyridinyl)-1-propanamine
Formel:
C₈H₁₂N₂ Molecular Weight: 136,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09802326 CAS-Nummer: 60289-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...