Methyl 3-pyridylacetate hydrochloride 95%

Lieferant: Apollo Scientific

OR965088-1G OR965088-5G
APOSOR965088-1GEA 168 EUR
APOSOR965088-1G APOSOR965088-5G
Methyl 3-pyridylacetate hydrochloride 95%
Methyl 3-pyridylacetate hydrochloride
Formel: C₈H₉NO₂·ClH
Molecular Weight: 187,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442777
CAS-Nummer: 69966-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR