2-Bromo-1-(4-chlorophenyl)propan-1-one 98%

Lieferant: Apollo Scientific
Achtung

OR965110-1G OR965110-5G
APOSOR965110-1GEA 171 EUR
APOSOR965110-1G APOSOR965110-5G
2-Bromo-1-(4-chlorophenyl)propan-1-one 98%
2-Bromo-1-(4-chlorophenyl)propan-1-one
Formel: C₉H₈BrClO
Molecular Weight: 247,516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018687
CAS-Nummer: 877-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR