3-Propyl-1H-pyrazole-5-carboxylic acid 96%
Lieferant: Apollo Scientific
OR965138-5G
APOSOR965138-5GEA
173
EUR
APOSOR965138-5G
3-Propyl-1H-pyrazole-5-carboxylic acid 96%
3-Propyl-1H-pyrazole-5-carboxylic acid
Formel:
C₇H₁₀N₂O₂ Molecular Weight: 154,167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02169458 CAS-Nummer: 76424-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...