1-(4-Hydroxyphenyl)-2-methoxyethanone 95%

Lieferant: Apollo Scientific

OR965180-1G
APOSOR965180-1GEA 30.6 EUR
APOSOR965180-1G
1-(4-Hydroxyphenyl)-2-methoxyethanone 95%
1-(4-Hydroxyphenyl)-2-methoxyethanone
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08461683
CAS-Nummer: 32136-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR