4-Pyridinebutanol 97%

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-4-yl)butan-1-ol

OR965183-5G
APOSOR965183-5GEA 98.6 EUR
APOSOR965183-5G
4-Pyridinebutanol 97%
4-Pyridinebutanol
Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265696
CAS-Nummer: 5264-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR