2-(4-Chlorophenyl)-3-oxobutanenitrile 96%
Lieferant: Apollo Scientific
OR965277-1G
OR965277-5G
OR965277-100MG
OR965277-250MG
OR965277-500MG
APOSOR965277-1GEA
258
EUR
APOSOR965277-1G
APOSOR965277-5G
APOSOR965277-100MG
APOSOR965277-250MG
APOSOR965277-500MG
2-(4-Chlorophenyl)-3-oxobutanenitrile 96%
2-(4-Chlorophenyl)-3-oxobutanenitrile
Formel:
C₁₀H₈ClNO Molecular Weight: 193,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01672899 CAS-Nummer: 5219-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...