5-(Fur-2-yl)thiophene-2-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9653-1G
OR9653-250MG
APOSOR9653-1GEA
235
EUR
APOSOR9653-1G
APOSOR9653-250MG
5-(Fur-2-yl)thiophene-2-carboxaldehyde 97%
5-(Fur-2-yl)thiophene-2-carboxaldehyde
Formel:
C₉H₆O₂S Molecular Weight: 178.208 g/mol Schmelzpunkt: 40…41.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410178 CAS-Nummer: 868755-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...