4-(1,2,3-Thiadiazol-4-yl)pyridine 95%

Lieferant: Apollo Scientific

OR965363-1G OR965363-250MG
APOSOR965363-1GEA 328 EUR
APOSOR965363-1G APOSOR965363-250MG
4-(1,2,3-Thiadiazol-4-yl)pyridine 95%
4-(1,2,3-Thiadiazol-4-yl)pyridine
Formel: C₇H₅N₃S
Molecular Weight: 163,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793696
CAS-Nummer: 102253-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR