4-(1H-1,2,3-Triazol-1-yl)aniline 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Amino-phenyl)-1,2,3-triazole

OR965381-1G OR965381-250MG
APOSOR965381-1GEA 148 EUR
APOSOR965381-1G APOSOR965381-250MG
4-(1H-1,2,3-Triazol-1-yl)aniline 95%
4-(1H-1,2,3-Triazol-1-yl)aniline
Formel: C₈H₈N₄
Molecular Weight: 160.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09048597
CAS-Nummer: 16279-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR