Cyclopropyl(pyridin-2-yl)methanamine 96%

Lieferant: Apollo Scientific

OR965497-1G OR965497-250MG OR965497-5G
APOSOR965497-1GEA 270 EUR
APOSOR965497-1G APOSOR965497-250MG APOSOR965497-5G
Cyclopropyl(pyridin-2-yl)methanamine 96%
Cyclopropyl(pyridin-2-yl)methanamine
Formel: C₉H₁₂N₂
Molecular Weight: 148.205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05191946
CAS-Nummer: 535925-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR