(S)-3-Methyl-pentanoic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(S)-3-Methylvaleric Acid
OR965509-1G
OR965509-5G
APOSOR965509-1GEA
78.3
EUR
APOSOR965509-1G
APOSOR965509-5G
(S)-3-Methyl-pentanoic acid 95%
(S)-3-Methyl-pentanoic acid
Formel:
C₆H₁₂O₂ Molecular Weight: 116,158 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02262177 CAS-Nummer: 1730-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...