4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
4-(((Benzyloxy)carbonyl)amino)-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid
OR965510-1G
OR965510-5G
APOSOR965510-1GEA
36.4
EUR
APOSOR965510-1G
APOSOR965510-5G
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid
Formel:
C₁₉H₂₆N₂O₆ Molecular Weight: 378.42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683056 CAS-Nummer: 288154-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...