3-Hydroxy-n,n-dimethylbenzamide 95%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-N,N-dimethylbenzamide

OR965513-1G OR965513-5G
APOSOR965513-1GEA 191 EUR
APOSOR965513-1G APOSOR965513-5G
3-Hydroxy-n,n-dimethylbenzamide 95%
3-Hydroxy-n,n-dimethylbenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01168918
CAS-Nummer: 15789-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR