4-(4-Cyanophenyl)piperidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-piperidin-4-ylbenzonitrile, 4-(Piperidin-4-yl)benzonitrile
OR965558-250MG
OR965558-1G
APOSOR965558-250MGEA
63.8
EUR
APOSOR965558-250MG
APOSOR965558-1G
4-(4-Cyanophenyl)piperidine
4-(4-Cyanophenyl)piperidine
Formel:
C₁₂H₁₄N₂ Molecular Weight: 186.253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426383 CAS-Nummer: 149554-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...