2-Chloro-1-(5-methyl-thiophen-2-yl)-ethanone 95%
Lieferant: Apollo Scientific
OR965590-100MG
OR965590-1G
OR965590-250MG
APOSOR965590-100MGEA
154
EUR
APOSOR965590-100MG
APOSOR965590-1G
APOSOR965590-250MG
2-Chloro-1-(5-methyl-thiophen-2-yl)-ethanone 95%
2-Chloro-1-(5-methyl-thiophen-2-yl)-ethanone
Formel:
C₇H₇ClOS Molecular Weight: 174,648 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464874 CAS-Nummer: 31772-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...