(2-Ethoxy-phenoxy)-acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Ethoxyphenoxy)acetic acid

OR965595-1G OR965595-5G
APOSOR965595-1GEA 104 EUR
APOSOR965595-1G APOSOR965595-5G
(2-Ethoxy-phenoxy)-acetic acid 95%
(2-Ethoxy-phenoxy)-acetic acid
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01124887
CAS-Nummer: 3251-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR