2-Chloro-4h-pyrido[1,2-a]pyrimidin-4-one 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-4H-pyrido[1,2-a]pyrimidin-4-one

OR965622-1G OR965622-250MG
APOSOR965622-1GEA 111 EUR
APOSOR965622-1G APOSOR965622-250MG
2-Chloro-4h-pyrido[1,2-a]pyrimidin-4-one 98%
2-Chloro-4h-pyrido[1,2-a]pyrimidin-4-one
Formel: C₈H₅ClN₂O
Molecular Weight: 180.591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00457859
CAS-Nummer: 5418-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR