Chlorosulphonyl acetic acid ethyl ester 97%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(chlorosulphonyl)acetate
OR965629-25G
OR965629-5G
APOSOR965629-25GEA
458
EUR
APOSOR965629-25G
APOSOR965629-5G
Chlorosulphonyl acetic acid ethyl ester 97%
Chlorosulphonyl acetic acid ethyl ester
Formel:
C₄H₇ClO₄S Molecular Weight: 186.614 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08458866 CAS-Nummer: 55896-93-0 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...