Methyl (chlorosulphonyl)acetate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 2-chlorosulphonylacetate, Methyl 2-(chlorosulphonyl)acetate
OR965635-1G
OR965635-5G
APOSOR965635-1GEA
256
EUR
APOSOR965635-1G
APOSOR965635-5G
Methyl (chlorosulphonyl)acetate
Methyl (chlorosulphonyl)acetate
Formel:
C₃H₅ClO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075558 CAS-Nummer: 56146-83-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...