3-Phenylamino-propionic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-(Phenylamino)propanoic acid

OR965638-1G OR965638-5G
APOSOR965638-1GEA 62.5 EUR
APOSOR965638-1G APOSOR965638-5G
3-Phenylamino-propionic acid 95%
3-Phenylamino-propionic acid
Formel: C₉H₁₁NO₂
Molecular Weight: 165,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089905
CAS-Nummer: 5652-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR