4-(4-Methylphenyl)piperidine 98%
Lieferant: Apollo Scientific
OR965671-1G
OR965671-5G
APOSOR965671-1GEA
190
EUR
APOSOR965671-1G
APOSOR965671-5G
4-(4-Methylphenyl)piperidine 98%
4-(4-Methylphenyl)piperidine
Formel:
C₁₂H₁₇N Molecular Weight: 175,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839928 CAS-Nummer: 59083-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...