6-(Piperidin-1-yl)pyridine-2-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(6-Formylpyridin-2-yl)piperidine, 2-Formyl-6-(piperidin-1-yl)pyridine
OR9657-1G
OR9657-250MG
APOSOR9657-1GEA
364
EUR
APOSOR9657-1G
APOSOR9657-250MG
6-(Piperidin-1-yl)pyridine-2-carboxaldehyde 97%
6-(Piperidin-1-yl)pyridine-2-carboxaldehyde
Formel:
C₁₁H₁₄N₂O Molecular Weight: 190,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08543443 CAS-Nummer: 859850-71-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...