4-Anilino-4-oxobutanoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 4-Oxo-4-(phenylamino)butanoic acid

OR965850-1G OR965850-5G
APOSOR965850-1GEA 37.7 EUR
APOSOR965850-1G APOSOR965850-5G
4-Anilino-4-oxobutanoic acid 95%
4-Anilino-4-oxobutanoic acid
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00029825
CAS-Nummer: 102-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR