1-Oxa-7-azaspiro[4.4]nonane hydrochloride

Lieferant: Apollo Scientific

OR966366-1G OR966366-250MG OR966366-500MG
APOSOR966366-1GEA 1180 EUR
APOSOR966366-1G APOSOR966366-250MG APOSOR966366-500MG
1-Oxa-7-azaspiro[4.4]nonane hydrochloride
1-Oxa-7-azaspiro[4.4]nonane hydrochloride
Formel: C₇H₁₃NO·ClH
Molecular Weight: 163,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28502507
CAS-Nummer: 1419590-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR