4-(Thien-2-yl)phenacyl bromide 97%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-4'-(thien-2-yl)acetophenone, 2-Bromo-1-[4-(thien-2-yl)phenyl]ethan-1-one

OR9681-1G OR9681-250MG
APOSOR9681-1GEA 212 EUR
APOSOR9681-1G APOSOR9681-250MG
4-(Thien-2-yl)phenacyl bromide 97%
4-(Thien-2-yl)phenacyl bromide
Formel: C₁₂H₉BrOS
Molecular Weight: 281,168 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06659059
CAS-Nummer: 128746-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR