4-(Thien-2-yl)benzyl bromide 97%

Lieferant: Apollo Scientific

OR9682-1G OR9682-5G
APOSOR9682-1GEA 361 EUR
APOSOR9682-1G APOSOR9682-5G
4-(Thien-2-yl)benzyl bromide 97%
4-(Thien-2-yl)benzyl bromide
Formel: C₁₁H₉BrS
Lagertemperatur: Raumtemperatur
MDL: MFCD06659060
CAS-Nummer: 81443-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR