Methyl 4-bromo-2-methylsulphanylbenzoate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-bromo-2-(methylthio)benzoate

OR968327-1G OR968327-500MG
APOSOR968327-1GEA 1150 EUR
APOSOR968327-1G APOSOR968327-500MG
Methyl 4-bromo-2-methylsulphanylbenzoate 97%
Methyl 4-bromo-2-methylsulphanylbenzoate
Formel: C₉H₉BrO₂S
Molecular Weight: 261,136 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22488927
CAS-Nummer: 1824127-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR