2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Carboxyphenyl)-2,5-dimethyl-1H-pyrrole
OR9731-1G
OR9731-250MG
APOSOR9731-1GEA
143
EUR
APOSOR9731-1G
APOSOR9731-250MG
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid 97%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
Formel:
C₁₃H₁₃NO₂ Molecular Weight: 215.248 g/mol Schmelzpunkt: 125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01125264 CAS-Nummer: 92028-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...