2,6-Dichloro-4-(trichloromethyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: α,α,α,2,6-Pentachloro-4-picoline

OR9768-25G OR9768-5G OR9768-100G
APOSOR9768-25GEA 333 EUR
APOSOR9768-25G APOSOR9768-5G APOSOR9768-100G
2,6-Dichloro-4-(trichloromethyl)pyridine 95%
2,6-Dichloro-4-(trichloromethyl)pyridine
Formel: C₆H₂Cl₅N
Molecular Weight: 265.352 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788280
CAS-Nummer: 22652-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR