2-(Morpholin-4-yl)isonicotinaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Morpholin-4-yl)pyridine-4-carboxaldehyde, 4-(4-Formylpyridin-2-yl)morpholine

OR9796-1G OR9796-250MG
APOSOR9796-1GEA 299 EUR
APOSOR9796-1G APOSOR9796-250MG
2-(Morpholin-4-yl)isonicotinaldehyde 97%
2-(Morpholin-4-yl)isonicotinaldehyde
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,214 g/mol
Schmelzpunkt: 62…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060524
CAS-Nummer: 864068-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR