3'-[(Pyrrolidin-1-yl)ethynyl]acetanilide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-{3-[(Pyrrolidin-1-yl)ethynyl]phenyl}acetamide

OR9862-1G OR9862-500MG
APOSOR9862-1GEA 364 EUR
APOSOR9862-1G APOSOR9862-500MG
3'-[(Pyrrolidin-1-yl)ethynyl]acetanilide 95%
3'-[(Pyrrolidin-1-yl)ethynyl]acetanilide
Formel: C₁₄H₁₆N₂O
Molecular Weight: 228,29 g/mol
Schmelzpunkt: 127…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659620
CAS-Nummer: 900014-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR