2-Methyl-6-phenylpyrimidin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-2-methyl-6-phenyl-1,3-diazine, 4-Hydroxy-2-methyl-6-phenylpyrimidine

OR9874-1G OR9874-250MG
APOSOR9874-1GEA 290 EUR
APOSOR9874-1G APOSOR9874-250MG
2-Methyl-6-phenylpyrimidin-4-ol
2-Methyl-6-phenylpyrimidin-4-ol
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00179595
CAS-Nummer: 62260-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR